Learn More
ATCC Genomic DNA from; Camelpox virus; Strain: V78-I-2379
Catalog No. 502384290
Encompass
Genomic DNA was isolated from a preparation of cell lysate and supernatant from Cercopithecus aethiops kidney epithelial cells infected with camelpox virus (CMLV) strain V78-I-2379. This product can be used in exclusivity testing for Mpox.
Catalog No. 50-238-4290 Supplier ATCC Supplier No. VR3375D
Genomic DNA was isolated from a preparation of cell lysate and supernatant from Cercopithecus aethiops kidney epithelial cells infected with camelpox virus (CMLV) strain V78-I-2379. This product can be used in exclusivity testing for Mpox.