Learn More
ATCC Genomic DNA from; Cowpox virus; Strain Brighton Red
Catalog No. 502382747
Encompass
Genomic DNA isolated from a preparation of Cowpox virus strain Brighton (ATCC VR-302). This product can be used for PCR and other molecular biology applications.
Catalog No. 50-238-2747 Supplier ATCC Supplier No. VR302D
Genomic DNA isolated from a preparation of Cowpox virus strain Brighton (ATCC VR-302). This product can be used for PCR and other molecular biology applications.