Learn More
ATCC Nostoc punctiforme
Supplier:  ATCC 29133
Encompass
Nostoc punctiforme strain PCC 73102 was isolated from Macrozamia sp. root in Australia. This cyanobacterium is known to produce glycolipid and ornithine carbamoyltransferase.
Catalog No. 50-238-1613