Learn More
ATCC Kocuria kristinae; Strain Vitek #12917
Supplier:  ATCC BAA752
Encompass
This organism is included in the bioMérieux VITEK 2 - GP ID Card Quality Control Organism Set.
Catalog No. 50-238-0802