Learn More
ATCC Genomic DNA from; Monkeypox virus; Isolate: hMPXV/USA/MA001/2022
Catalog No. 502383614
Encompass
Quantitative Genomic DNA from Monkeypox virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3614 Supplier ATCC Supplier No. VR3371D
Quantitative Genomic DNA from Monkeypox virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.