Learn More
ATCC Rhizobium radiobacter
Catalog No. 502380577
Encompass
Rhizobium radiobacter strain K1026 was derived from strain K84 (ATCC 49644). This strain contains the plasmid pAgK1026 and has applications in the biological control of crown gall.
Catalog No. 50-238-0577 Supplier ATCC Supplier No. 700691
Rhizobium radiobacter strain K1026 was derived from strain K84 (ATCC 49644). This strain contains the plasmid pAgK1026 and has applications in the biological control of crown gall.