Learn More
Biochain Institute Inc Genomic DNA - Human Diabetic Diseased Tissue: Small Intestine: Duodenum, from a single donor, 50 ug/PK
SDP
Catalog No. 50181419
Encompass_Preferred
BioChain's tissue genomic DNAs are isolated from a wide variety of tissues using a proprietary modified guanidine thiocyanate technique. The quality and purity of genomic DNA are tested by spectrophotometer and electrophoresis. A260/280 is between 1.8 and 2.0, and A260/230 is >2.0 (detected in 10 mM Tris-Cl, pH 7.5). Contamination of RNA, polysaccharides and proteoglycans has been effectively eliminated.
Catalog No. 50-181-419 Supplier Biochain Institute Inc Supplier No. D1236101DIA
BioChain's tissue genomic DNAs are isolated from a wide variety of tissues using a proprietary modified guanidine thiocyanate technique. The quality and purity of genomic DNA are tested by spectrophotometer and electrophoresis. A260/280 is between 1.8 and 2.0, and A260/230 is >2.0 (detected in 10 mM Tris-Cl, pH 7.5). Contamination of RNA, polysaccharides and proteoglycans has been effectively eliminated.