Learn More
Norgen Biotek Corp Genomic DNA Isolation Kit, 100 Prep
Catalog No. NC1522502
Encompass_Preferred
This kit is designed for the rapid preparation of genomic DNA from various tissue samples, cultured cells, viruses, bodily fluids and swabs using a rapid spin column protocol. Purified DNA is of an excellent yield and quality, and is immediately ready for any downstream application including PCR, qPCR, genotyping, sequencing and more. The protocol can be completed in approximately 80 minutes (including incubation time).
Catalog No. NC1522502 Supplier Norgen Biotek Corp Supplier No. 24750
This kit is designed for the rapid preparation of genomic DNA from various tissue samples, cultured cells, viruses, bodily fluids and swabs using a rapid spin column protocol. Purified DNA is of an excellent yield and quality, and is immediately ready for any downstream application including PCR, qPCR, genotyping, sequencing and more. The protocol can be completed in approximately 80 minutes (including incubation time).