Learn More
LUNA NANOTECH INC Genomic Dna Purification Kit, 50 Preps
Supplier:  LUNA NANOTECH INC NK06150
Encompass_Preferred
PuroSPIN™ Genomic DNA Purification Kit, 50 Preps
PuroSPIN™ Genomic DNA Purification Kitis designed to provide simple and easy-to-use method for extraction and purification of genomic DNA from Gram-positive and Gram-negative bacteria, yeast, mammalian cells, whole blood and animal tissues.
The purification technology is based on nucleic acid binding to silica spin-columns, which allows efficient removal of protein, salt and other contaminants. Silica spin columns offer faster, simpler and safer alternative to traditional nucleic acid purification techniques such as phenol-chloroform extraction.
PuroSPIN™ buffer formulations produce highly purified DNA product for downstream applications, such as sequencing, ligation, transfection, Southern blotting, qPCR, or microarray analysis.
PuroSPIN™ columns can bind up to 200 μg of DNA product.
Features:
- Easily purify genomic DNA using
Catalog No. 50-256-6953