Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies Genomic DNA ScreenTape, 200 to > 60,000 bp, 105 Samples

Catalog No. NC1784723
Encompass

This genomic DNA ScreenTape assay is designed for automated electrophoretic separation, sizing, quantification, and integrity analysis of genomic DNA samples. It is compatible with all TapeStation systems and enables the calculation of the DNA Integrity Number (DIN) as a quality metric, providing a standardized approach for sample QC, including for NGS workflows.

  • Sizing range: 200 bp to >60,000 bp
  • Quantification range: 10 ng/μL to 100 ng/μL
  • Sensitivity: 0.5 ng/μL
  • Sample volume required: 1 μL
  • Quantitative accuracy: ±20 %
  • Quantitative precision: 15 % CV
  • Sizing accuracy: ±15 %
  • Sizing precision: 15 % CV
  • Application: Suitable for diverse sample sources, including FFPE material
  • Analysis time: <25 minutes for 15 samples

Catalog No. NC1784723 Supplier Agilent Technologies Supplier No. 50675365
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This genomic DNA ScreenTape assay is designed for automated electrophoretic separation, sizing, quantification, and integrity analysis of genomic DNA samples. It is compatible with all TapeStation systems and enables the calculation of the DNA Integrity Number (DIN) as a quality metric, providing a standardized approach for sample QC, including for NGS workflows.

  • Sizing range: 200 bp to >60,000 bp
  • Quantification range: 10 ng/μL to 100 ng/μL
  • Sensitivity: 0.5 ng/μL
  • Sample volume required: 1 μL
  • Quantitative accuracy: ±20 %
  • Quantitative precision: 15 % CV
  • Sizing accuracy: ±15 %
  • Sizing precision: 15 % CV
  • Application: Suitable for diverse sample sources, including FFPE material
  • Analysis time: <25 minutes for 15 samples

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.