Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zymo Research Corporation Genomic Lysis Buffer, 200 ml
SDP

Catalog No. 502037027
Encompass_Preferred

Genomic Lysis Buffer 2 x 100 ml

Catalog No. 50-203-7027 Supplier Zymo Research Corporation Supplier No. D30041200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Genomic Lysis Buffer 2 x 100 ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.