Learn More
ATCC Genomic RNA from SARS-CoV-2; Variant B.1.351; Strain: USA/MD-HP01542/2021
Supplier:  ATCC VR3327D
Encompass
Genomic RNA isolated from a preparation of Severe acute respiratory syndrome-related coronavirus 2 isolate USA/MD-HP01542/2021. This product can be used in assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays.
Catalog No. 50-238-3407