Learn More
ATCC Quantitative genomic RNA from Severe acute respiratory syndrome-related coronavirus 2 strain 2019-nCoV/USA-WA1/2020 (ATCC VR-1986D™)
Catalog No. 502383074
Encompass
Genomic RNA isolated from a preparation of Severe acute respiratory syndrome-related coronavirus 2 strain 2019-nCoV/USA-WA1/2020. It is used for assay development, verification, and qPCR standard curve generation. Recommended for molecular diagnostics and respiratory disease research.
Catalog No. 50-238-3074 Supplier ATCC Supplier No. VR1986D
Genomic RNA isolated from a preparation of Severe acute respiratory syndrome-related coronavirus 2 strain 2019-nCoV/USA-WA1/2020. It is used for assay development, verification, and qPCR standard curve generation. Recommended for molecular diagnostics and respiratory disease research.