Learn More
ATCC Genomic RNA from; SARS-CoV-2 Strain:; Germany/BavPat1/2020
Catalog No. 502384524
Encompass
Quantitative genomic RNA from Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2) strain Germany/BavPat1/2020 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4524 Supplier ATCC Supplier No. VR1994D
Quantitative genomic RNA from Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2) strain Germany/BavPat1/2020 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.