Learn More
ATCC Genomic RNA from; SARS-CoV-2; Omicron, BA.5; Strain: USA/COR-22-063113/2022
Supplier:  ATCC VR3378D
Encompass
Quantitative genomic RNA from Severe acute respiratory syndrome-related coronavirus 2 strain USA/COR-22-063113/2022 (Omicron variant, BA.5) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2418