Learn More
ATCC Genomic RNA from; SARS-CoV-2; variant B.1.1.7
Catalog No. 502384212
Encompass
Genomic RNA isolated from a preparation of Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2) isolate USA/CA_CDC_5574/2020. This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4212 Supplier ATCC Supplier No. VR3326D
Genomic RNA isolated from a preparation of Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2) isolate USA/CA_CDC_5574/2020. This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.