Learn More
Sigma Aldrich Fine Chemicals Biosciences GenomiphiTM V2 DNA Amplifi
Catalog No. 502908815
Encompass
Illustra GenomiPhi(TM) V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 mug yield from nanogram amounts of DNA sample.illustra(TM) GenomiPhi(TM) V2 DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva. It contains all of the components necessary for mini-scale whole genome amplification by isothermal strand displacement amplification.Amplification is highly uniform over the entire genome so that locus representation remains extremely close to the original DNA sample. Furthermore amplification is carried out with very high fidelity due to Phi29 DNA polymerase proofreading activity.The illustra(TM) GenomiPhi(TM) V2 DNA Amplification Kit procedure is quick simple and does not require a thermocycler. A typical DNA yield of 4 to 7 mug DNA can be achieved in less than two hours with little hands-on time. The average product length is over 10 kb. The starting material for GenomiPhi(TM) reactions ca
Catalog No. 50-290-8815 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE25660032
Illustra GenomiPhi(TM) V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 mug yield from nanogram amounts of DNA sample.illustra(TM) GenomiPhi(TM) V2 DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva. It contains all of the components necessary for mini-scale whole genome amplification by isothermal strand displacement amplification.Amplification is highly uniform over the entire genome so that locus representation remains extremely close to the original DNA sample. Furthermore amplification is carried out with very high fidelity due to Phi29 DNA polymerase proofreading activity.The illustra(TM) GenomiPhi(TM) V2 DNA Amplification Kit procedure is quick simple and does not require a thermocycler. A typical DNA yield of 4 to 7 mug DNA can be achieved in less than two hours with little hands-on time. The average product length is over 10 kb. The starting material for GenomiPhi(TM) reactions ca