Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GenoTube Livestock Swabs

Catalog No. 9062010
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 piece
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-620-10 1 piece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-620-10 Supplier Thermo Scientific™ Supplier No. 9062010
  • $4.35
    $3.65 / Each of 1

    Save $0.70

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems™ GenoTube Livestock Swab is easy to use and can be handled by even novice users.

With its proprietary drying system, GenoTube Livestock Swabs rapidly reduce enzyme activity, preventing the breakdown of nucleic acids and proteins that are essential for many laboratory diagnostic procedures. Once dried, collected samples can be stored without refrigeration for simple and cost-effective transportation. In the lab, the samples can be easily reconstituted and used for numerous analyses.

  • Excellent sample quality—fast drying, not influenced by local humidity
  • Convenient versatility—capable of collecting different sample matrices, such as nasal swabs, oral fluids, blood, and fecal samples
  • Cost-effective—low transportation costs because refrigeration or rapid transport not required
  • Individually packaged—allows a unique identifier on both the swab cap and tube for clear identification and also reduces the potential for cross-contamination
  • Superior conservation—no degradation of DNA after long-term storage
  • Sample flexibility—sample can be used for antigen or antibody diagnostics (e.g., PCR, ELISA, etc.)

Genotyping applications

Collecting DNA samples for genotyping and sequencing using the GenoTube Livestock Swab is easy and rapid. After the swab containing the sample is placed back in the tube, the sample is dried, and the DNA in the sample is conserved. The tube can be transported and stored under normal conditions without refrigeration. DNA contained in the sample remains stable for years.

NOTE: this product has a minimum order quantity of 100 pieces.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Prionics
Content And Storage 1 GenoTube Livestock Swab

Store at 15–30°C.

Shipping Condition Room Temperature
Quantity 1 piece
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.