Learn More
Apexbio Technology LLC Genotyping Kit, 500 rxns. Cas: MFCD: NA. Genotyping Kit for target alleles of insects, tissues, fishes and cells
SDP
Catalog No. 501905572
Encompass_Preferred
The Genotyping Kit for target alleles of insects, tissues, fishes and cells provides convenience in faster preparation and superior PCR amplification. Lysis buffer and balance buffer could digest tissues or cells rapidly to release unbroken genomic DNA, and it could be used as PCR template directly without extraction. There is no need for overnight digestion, Phenol/Chloroform extraction, toilsome manual purification or the utilization of expensive DNA spin columns. By using our kit, you can substantially shorten your time for digestion. Furthermore, the 2PCR Master Mix (With Dye) will guarantee a robust and accurate amplification consequence. This Master Mix contains dye which means it can be directly electrophoresed after the amplification without the need to add a loading buffer. In addition, our genotyping Kit mitigates the risk of sample cross-contamination occurring in the PCR reaction by conducting the DNA extraction in one single tube. Other sizes are available.
Catalog No. 50-190-5572 Supplier Apexbio Technology LLC Supplier No. K1026500
The Genotyping Kit for target alleles of insects, tissues, fishes and cells provides convenience in faster preparation and superior PCR amplification. Lysis buffer and balance buffer could digest tissues or cells rapidly to release unbroken genomic DNA, and it could be used as PCR template directly without extraction. There is no need for overnight digestion, Phenol/Chloroform extraction, toilsome manual purification or the utilization of expensive DNA spin columns. By using our kit, you can substantially shorten your time for digestion. Furthermore, the 2PCR Master Mix (With Dye) will guarantee a robust and accurate amplification consequence. This Master Mix contains dye which means it can be directly electrophoresed after the amplification without the need to add a loading buffer. In addition, our genotyping Kit mitigates the risk of sample cross-contamination occurring in the PCR reaction by conducting the DNA extraction in one single tube. Other sizes are available.