Learn More
Cole-Parmer Jenway Genova Plus Life Science Spectrophotometer, DNA/RNA/Oligo/Protein; 115-230 VAC
Catalog No. 501961223 Shop All Cole-Parmer Products
Encompass_Preferred
The Genova Plus is a UV/visible spectrophotometer dedicated to life science analysis. This versatile spectrophotometer can also be used as a standard spectrophotometer. It allows the measurement of DNA concentrations and purity ratios using wavelengths recorded at 260, 280 and 230nm, with an optional correction at 320nm.The Genova Plus is pre-programmed with Bradford, Lowry, Biuret, BCA and Direct UV methods for protein analysis and features an OD measurement mode enabling users to measure optical density at 600nm for cell harvesting.
Catalog No. 50-196-1223 Supplier Cole-Parmer Supplier No. 8307000
Add to Cart
Add to Cart
Request a Quote
The Genova Plus is a UV/visible spectrophotometer dedicated to life science analysis. This versatile spectrophotometer can also be used as a standard spectrophotometer. It allows the measurement of DNA concentrations and purity ratios using wavelengths recorded at 260, 280 and 230nm, with an optional correction at 320nm.The Genova Plus is pre-programmed with Bradford, Lowry, Biuret, BCA and Direct UV methods for protein analysis and features an OD measurement mode enabling users to measure optical density at 600nm for cell harvesting.