Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Gentamicin 10 mg/mL, Pack 5 X 10 mL
SDP

Catalog No. 50983231
Encompass_Preferred

Gentamicin is an aminoglycoside complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is used as the sulfate salt. Gentamicin sulfate is a broad-spectrum antibiotic used as a selection agent (gentamicin-resistance gene) in molecular biology and cell culture applications.

This product is formulated to contain 10 mg/mL Gentamicin in deionized water Recommended for use in eukaryotic cell culture at a volume of 1 milliliter per liter.

Ready to use solution

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 36 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-231 Supplier Quality Biological Inc Supplier No. 120099661
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gentamicin is an aminoglycoside complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is used as the sulfate salt. Gentamicin sulfate is a broad-spectrum antibiotic used as a selection agent (gentamicin-resistance gene) in molecular biology and cell culture applications.

This product is formulated to contain 10 mg/mL Gentamicin in deionized water Recommended for use in eukaryotic cell culture at a volume of 1 milliliter per liter.

Ready to use solution

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 36 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.