Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Gentamicin (10 mg/mL)

Catalog No. 15710064
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-710-064 10 mL
15-710-072 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-710-064 Supplier Gibco™ Supplier No. 15710064
  • $22.64
    $21.65 / Each of 1

    Save $0.99

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea

Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria.

  • Effective against wide variety of gram-positive and gram-negative bacteria
  • Used for prevention of bacterial contamination on cell cultures
  • Recommended working concentration ranges from 0.5 to 50μg/mL

Cell Culture, Mammalian Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Concentration 10 mg/mL
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
For Use With (Application) Prevention of Cell Culture Contamination
Quantity 10 mL
Shipping Condition Room Temperature
If Gentamicin (10 mg/mL) is accidentally stored at 2-8 degrees C, would it affect the stability of the antibiotic?

No, storing Gentamicin solution for several days at 2-8 degrees C will not have any negative impact on its performance or stability. However, as Gentamicin solution has been shown to be stable at room temperature, the recommended storage temperature is ~25 degrees C.

How can I decontaminate my cultures?

When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.

1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.

The following is a suggested procedure for determining toxicity levels and decontaminating cultures:

1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.

What antibiotics do you offer to help control or eliminate cell culture contamination?

Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.