Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Gentamicin (10 mg/mL)

Catalog No. p-9056013
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-710-064 10 mL
15-710-072 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-710-064 Supplier Gibco™ Supplier No. 15710064
Only null left
Add to Cart
Edge
Add to Cart

Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea

Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria.

  • Effective against wide variety of gram-positive and gram-negative bacteria
  • Used for prevention of bacterial contamination on cell cultures
  • Recommended working concentration ranges from 0.5 to 50μg/mL

Cell Culture, Mammalian Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Concentration 10 mg/mL
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
For Use With (Application) Prevention of Cell Culture Contamination
Quantity 10 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.