Learn More
Selleck Chemical LLC Gentamicin
SDP
Catalog No. 5000003686
Encompass_Preferred
Gentamicin an aminoglycoside class of bactericidal antibiotic is effective against gram-negative bacterial infections Gentamicin can be used to induce animal models of Renal Failure
Catalog No. 50-000-03686 Supplier Selleck Chemical LLC Supplier No. S628925MG
Gentamicin an aminoglycoside class of bactericidal antibiotic is effective against gram-negative bacterial infections Gentamicin can be used to induce animal models of Renal Failure