Gentamicin is an aminoglycoside complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is used as the sulfate salt. Gentamicin sulfate is a broad-spectrum antibiotic used as a selection agent (gentamicin-resistance gene) in molecular biology and cell culture applications.
This product is formulated to contain 50 mg/mL Gentamicin in deionized water Recommended for use in eukaryotic cell culture at a volume of 1 milliliter per liter.
Ready to use solution
Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 36 months from date of manufacture
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.