Learn More
ILEX LIFE SCIENCES LLC GENTAMICIN 50MG/ML 100 ML
SDP
Catalog No. 502733731
Encompass_Preferred
PAN-Biotech Gentamicin Sulfate Solution for Cell Culture, 50 mg/ml (100 ml). COMPOSITION: Gentamicin sulfate: 50.00 g/L; Sodium chloride: 8.50 g/L. DESCRIPTION: Gentamicin is an aminoglycoside antibiotic produced by Micromonospora. Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria. APPLICATION: Gentamicin Sulfate is often used as an antibiotic agent to prevent bacterial contamination of cell cultures. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP.: -20°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. USAGE: This product is intended for Laboratory Research Use Only. Not for use in diagnostic or therapeutic procedures.
Catalog No. 50-273-3731 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0613100
PAN-Biotech Gentamicin Sulfate Solution for Cell Culture, 50 mg/ml (100 ml). COMPOSITION: Gentamicin sulfate: 50.00 g/L; Sodium chloride: 8.50 g/L. DESCRIPTION: Gentamicin is an aminoglycoside antibiotic produced by Micromonospora. Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria. APPLICATION: Gentamicin Sulfate is often used as an antibiotic agent to prevent bacterial contamination of cell cultures. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP.: -20°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. USAGE: This product is intended for Laboratory Research Use Only. Not for use in diagnostic or therapeutic procedures.