Learn More
ACROBiosystems - Gentamicin 96tests
Catalog No. 502534246
Encompass_Preferred
Residues of gentamicin are prone to occur in the production process of biological products which can easily lead to abnormal reactions in the human body. Therefore the residual amount of gentamicin in biological products or semi-finished products of biological products should be strictly controlled.
Catalog No. 50-253-4246 Supplier ACROBiosystems Supplier No. RESA02596TESTS
Residues of gentamicin are prone to occur in the production process of biological products which can easily lead to abnormal reactions in the human body. Therefore the residual amount of gentamicin in biological products or semi-finished products of biological products should be strictly controlled.