Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Gentamicin/Amphotericin Solution

Catalog No. 502722960
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-2960 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-272-2960 Supplier Gibco™ Supplier No. R01510
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Gentamicin/Amphotericin Solution

A sterile, concentrated (500x) antibiotic/antimycotic solution of gentamicin and amphotericin B (10 x 1 ml vials/pkg). One Gentamicin/Amphotericin Solution 10-Pak is enough to supplement ten 500 mL bottles of basal medium. Animal origin-free.

TRUSTED_SUSTAINABILITY

Specifications

Type Amphotericin B, Gentamicin
Physical Form Liquid
Sterile Filtered Yes
Concentration 500 X, 125 μg/mL Amphotericin B, 5 mg/mL Gentamicin
Quantity 10 x 1 mL
Validated Application Prevention of Cell Culture Contamination
Product Line Cascade Biologics
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.