Learn More
Medchemexpress LLC Gentamicin C1a | 26098-04-4 | 98.0% | 449.54 | 5 MG
SDP
Catalog No. 5000362316
Encompass_Preferred
Gentamicin C1a is a precursor of the semi-synthetic antibiotic Etimicin, exhibiting antibacterial activity. It is the major component of the Gentamicin complex.
  • Exhibits antibacterial activity
  • Inhibits *Escherichia coli*, *P. aeruginosa*, and *S. aureus* with an IC50 value of 1 mg/mL
  • Solid appearance, white to light yellow in color
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-62316 Supplier Medchemexpress LLC Supplier No. HY1482865MG
Gentamicin C1a is a precursor of the semi-synthetic antibiotic Etimicin, exhibiting antibacterial activity. It is the major component of the Gentamicin complex.
  • Exhibits antibacterial activity
  • Inhibits *Escherichia coli*, *P. aeruginosa*, and *S. aureus* with an IC50 value of 1 mg/mL
  • Solid appearance, white to light yellow in color
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light