Learn More
Cayman Chemical Gentamicin ELISA Monoclonal Antibody (6X), 1 mL
SDP
Catalog No. 502942464
Encompass_Preferred
Gentamicin ELISA monoclonal antibody (6X) has been tested and formulated to work exclusively with Cayman’s Gentamicin ELISA Kit (Item No. 502400).
Catalog No. 50-294-2464 Supplier Cayman Chemical Supplier No. 4005601 ML
Gentamicin ELISA monoclonal antibody (6X) has been tested and formulated to work exclusively with Cayman’s Gentamicin ELISA Kit (Item No. 502400).