Learn More
Cayman Chemical GentamIcIn-HRP Tracer6X 1mL
SDP
Catalog No. 502942331
Encompass_Preferred
Gentamicin-HRP tracer (6X) has been tested and formulated to work exclusively with Cayman’s Gentamicin ELISA Kit (Item No. 502400).
Catalog No. 50-294-2331 Supplier Cayman Chemical Supplier No. 4005591 ML
Gentamicin-HRP tracer (6X) has been tested and formulated to work exclusively with Cayman’s Gentamicin ELISA Kit (Item No. 502400).