Learn More
U.S. Pharmacopeia Gentamicin sulfate, 1405-41-0, MFCD00270181, 200 mg
Catalog No. NC1001426
Encompass
Gentamicin sulfate, a broad-spectrum antibiotic, used as a selection agent (gentamicin-resistance gene) in cell culture and molecular biology applications. This product is recommended for use at 50 mg/L. Synonym: Gentamicin sulfate salt.
Catalog No. NC1001426 Supplier U.S. Pharmacopeia Supplier No. 1289003
Gentamicin sulfate, a broad-spectrum antibiotic, used as a selection agent (gentamicin-resistance gene) in cell culture and molecular biology applications. This product is recommended for use at 50 mg/L. Synonym: Gentamicin sulfate salt.