Learn More
Medchemexpress LLC Gentamicin (sulfate) | 1405-41-0 | C24H55N7O11S3* | 5 G
SDP
Catalog No. 5000432277
Encompass_Preferred
Gentamicin sulfate is an aminoglycoside antibiotic that effectively inhibits the growth of both gram-positive and gram-negative bacteria, as well as several strains of mycoplasma in tissue culture. It has also been shown to inhibit DNase I with an IC50 of 0.57 mM.
  • Broad-spectrum antibiotic: inhibits both gram-positive and gram-negative bacteria.
  • Effective against mycoplasma: inhibits several strains of mycoplasma in tissue culture.
  • DNase I inhibitor: inhibits DNase I with an IC50 of 0.57 mM.
  • Non-toxic to tissue culture monolayers: does not inhibit virus replication and is non-toxic to Rhesus monkey kidney, HeLa, and human amnion cells.
  • Stable at autoclave temperatures: remains stable under autoclaving conditions.
  • Improved survival in sepsis: improves survival and alters the inflammatory profile in a murine model of sepsis from *P. aeruginosa*.
Catalog No. 50-004-32277 Supplier Medchemexpress LLC Supplier No. HYA02765G
Gentamicin sulfate is an aminoglycoside antibiotic that effectively inhibits the growth of both gram-positive and gram-negative bacteria, as well as several strains of mycoplasma in tissue culture. It has also been shown to inhibit DNase I with an IC50 of 0.57 mM.
  • Broad-spectrum antibiotic: inhibits both gram-positive and gram-negative bacteria.
  • Effective against mycoplasma: inhibits several strains of mycoplasma in tissue culture.
  • DNase I inhibitor: inhibits DNase I with an IC50 of 0.57 mM.
  • Non-toxic to tissue culture monolayers: does not inhibit virus replication and is non-toxic to Rhesus monkey kidney, HeLa, and human amnion cells.
  • Stable at autoclave temperatures: remains stable under autoclaving conditions.
  • Improved survival in sepsis: improves survival and alters the inflammatory profile in a murine model of sepsis from *P. aeruginosa*.