Learn More
Medchemexpress LLC Gentamicin (sulfate) | 1405-41-0 | MFCD00270181 | >80% | C24H55N7O11S3* | 1 G
SDP
Catalog No. 5000433441
Encompass_Preferred
Gentamicin sulfate is an aminoglycoside antibiotic that inhibits the growth of both gram-positive and gram-negative bacteria, and it also inhibits several strains of mycoplasma in tissue culture. It has been observed to inhibit DNase I with an IC50 of 0.57 mM.
  • Inhibits gram-positive and gram-negative bacteria
  • Inhibits mycoplasma in tissue culture
  • More effective in vitro bacterial inhibitor than penicillin-streptomycin
  • Nontoxic to tissue culture monolayers and does not inhibit virus replication
  • Used as an additive in commercial mycology media
  • Bactericidal for a wide range of organisms
  • Stable at autoclave temperatures
  • Improves survival and alters inflammatory profile in murine sepsis model
  • Down-regulates proinflammatory cytokines, IL-6
Catalog No. 50-004-33441 Supplier Medchemexpress LLC Supplier No. HYA02761G
Gentamicin sulfate is an aminoglycoside antibiotic that inhibits the growth of both gram-positive and gram-negative bacteria, and it also inhibits several strains of mycoplasma in tissue culture. It has been observed to inhibit DNase I with an IC50 of 0.57 mM.
  • Inhibits gram-positive and gram-negative bacteria
  • Inhibits mycoplasma in tissue culture
  • More effective in vitro bacterial inhibitor than penicillin-streptomycin
  • Nontoxic to tissue culture monolayers and does not inhibit virus replication
  • Used as an additive in commercial mycology media
  • Bactericidal for a wide range of organisms
  • Stable at autoclave temperatures
  • Improves survival and alters inflammatory profile in murine sepsis model
  • Down-regulates proinflammatory cytokines, IL-6