Learn More
Description
Gentamicin is an aminoglycoside antibiotic complex that inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus. It is effective against mycoplasma as well.
- Used as a selection agent (gentamicin-resistance gene) in molecular biology applications.
- Key applications: cell culture applications, antimicrobial treatment.
Specifications
Specifications
| Physical Form | Liquid |
| CAS | 1405-41-0, 7732-18-5 |
| pH | 4 to 6 |
| Sterility | Sterile |
| Color | Clear Colorless to Pale Yellow |
| Concentration | 50 mg/mL |
| Quantity | 1 x 10 mL |
| Synonym | Garamycin, Gentamicin sulfate salt, Gentiomycin C |
| Grade | Not Available |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.