Learn More
Enzo Life Sciences Gentamicin sulfate (5g). CAS: 1405-41-0
SDP
Supplier:  Enzo Life Sciences ALX380003G005
Encompass_Preferred
Broad-spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial and antimycoplasmal concentrations. Due to its extended stability and slow development of bacterial resistance, it is a useful antibiotic in long-term virus und tissue culture studies. Appearance: White to off-white powder. Solubility: Soluble in water; almost insoluble in alcohol or ether. Long Term Storage: +4°C.
Catalog No. 50-200-8898