Learn More
Selleck Chemical LLC Gentamicin Sulfate S4030-1g
SDP
Catalog No. 503109550
Encompass_Preferred
Gentamycin Sulfate (NSC-82261 SCH9724) is a broad-spectrum aminoglycoside antibiotic used for cell culture which inhibits protein synthesis in sensitive organisms Gentamicin Sulfate can be used to induce animal models of Renal Failure
Catalog No. 50-310-9550 Supplier Selleck Chemical LLC Supplier No. S40301G
Gentamycin Sulfate (NSC-82261 SCH9724) is a broad-spectrum aminoglycoside antibiotic used for cell culture which inhibits protein synthesis in sensitive organisms Gentamicin Sulfate can be used to induce animal models of Renal Failure