Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentamicin Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013056 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3056 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3056 Supplier MilliporeSigma Supelco Supplier No. PHR10771G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gentamicin Sulfate is a broad-spectrum aminoglycoside antibiotic, produced from Micromonospora purpura. It is widely used as an active pharmaceutical ingredient (API) with therapeutic potential against both Gram-positive and Gram-negative bacteria.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 1405-41-0
Physical Form Neat
MDL Number MFCD00270181
Synonym Gentamicin sulfate salt
Type Certified Reference Material
Quantity 1 g
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.