Learn More
Sigma Aldrich Fine Chemicals Biosciences Gentamicin sulfate salt po100G
Catalog No. 502908212
Encompass
Gentamicin also known as gentiomycin C is an aminoglycoside which is naturally produced by Gram-negative bacteria. This antibiotic functions by preventing protein synthesis. Gentamicin sulfate prevents the growth of both Gram positive and Gram negative bacteria in vivo and in vitro. In tissue culture gentamicin sulfate is used to inhibit the growth of various strains of mycoplasma.
Catalog No. 50-290-8212 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G1264100G
Add to Cart
Add to Cart
Request a Quote
Gentamicin also known as gentiomycin C is an aminoglycoside which is naturally produced by Gram-negative bacteria. This antibiotic functions by preventing protein synthesis. Gentamicin sulfate prevents the growth of both Gram positive and Gram negative bacteria in vivo and in vitro. In tissue culture gentamicin sulfate is used to inhibit the growth of various strains of mycoplasma.