Learn More
AdipoGen Gentamicin sulfate (USP Grade) (25 g)
SDP
Supplier:  AdipoGen AGCN20066G025
Encompass_Preferred
Chemical. CAS: 1405-41-0. Formula: C21H43N5O7 . H2SO4 (unspecified). MW: 575.6. Isolated from Micromonospora sp. Aminoglycoside antibiotic. Protein synthesis inhibitor. Causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site. Antibacterial against Gram-negative aerobic bacteria, Gram-positive bacteria and mycoplasmas. Used as a selection agent (gentamicin-resistance gene) in molecular biology applications. Broad-spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial and antimycoplasmal concentrations. Due to its extended stability and slow development of bacterial resistance, it is a useful antibiotic in long-term virus und tissue culture studies. Bactericidal effects are exerted by the binding to the outer membrane, causing disruption of the membrane.
Catalog No. 50-205-3412