Learn More
Gold Biotechnology Inc Gentamicin Sulfate, USP Grade 100 g
SDP
Catalog No. 501532781
Encompass_Preferred
Gentamicin is a broad-spectrum aminoglycoside antibiotic effective against gram-negative and gram-positive species. In addition, gentamicin exhibits antimicrobial properties against species resistant to tetracycline, colistin, kanamycin, and chloramphenicol.
Catalog No. 50-153-2781 Supplier Gold Biotechnology Inc Supplier No. G400100
Gentamicin is a broad-spectrum aminoglycoside antibiotic effective against gram-negative and gram-positive species. In addition, gentamicin exhibits antimicrobial properties against species resistant to tetracycline, colistin, kanamycin, and chloramphenicol.