Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentamicin Sulfate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S162502
Change view
Click to view available options
Quantity:
500 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162502 1000 g
S162508 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S162502 Supplier G-Biosciences Supplier No. RC859
Add to Cart
Add to Cart

Gentamicin efficiently inhibits growth of many Gram-positive and Gram-negative microorganisms.

It inhibits protein synthesis by binding to the L6 protein of the 30S ribosomal subunit. It is also effective against mycoplasma. Recommended for use in cell culture applications.

Chemical Identifiers

Molecular Weight (g/mol) 575.67 g/mol

Specifications

Boiling Point 797.6°C
Quantity 1000 g
Percent Purity 98%
Grade USP
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.