Learn More
Indofine Chemical Gentamicin Sulphat, 90%, 10 Gm
SDP
Catalog No. 50909893
Encompass_Preferred
Molecular Formula C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S, Purity: >90%, Molecular Weight 1488.790, Appearance: White to off-white powder, Storage conditions: Store at 0-8C, Also known as Gentamycin sulfate
Catalog No. 50-909-893 Supplier Indofine Chemical Supplier No. G0124 10 GM
Molecular Formula C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S, Purity: >90%, Molecular Weight 1488.790, Appearance: White to off-white powder, Storage conditions: Store at 0-8C, Also known as Gentamycin sulfate