Learn More
Crescent Chemical Co Inc GENTAMYCIN SOLUTION 50 MG/ML
SDP
Catalog No. 50249145
Encompass_Preferred
Gentamycin Solution 50 mg/ml sterile filtered Formulated to contain 50 mg/ml gentamycin base in deionized water. 50 mg gentamycin base correspond to approx. 50 000 units of gentamycin and are approx. 80 mg gentamycin sulfate. Size - 20ML Storage Conditions - -15 °C T
Catalog No. 50-249-145 Supplier Crescent Chemical Co Inc Supplier No. SE47991.01
Gentamycin Solution 50 mg/ml sterile filtered Formulated to contain 50 mg/ml gentamycin base in deionized water. 50 mg gentamycin base correspond to approx. 50 000 units of gentamycin and are approx. 80 mg gentamycin sulfate. Size - 20ML Storage Conditions - -15 °C T