Learn More
IBI SCIENTIFIC 3P Gentamycin Sulfate Solution | 1405-41-0 | MFCD00270181 | 20 mL
Catalog No. NC0753440
Encompass_Preferred
Gentamycin Sulfate Solution - 20ml Gentamycin Sulfate Solution is used as an inhibitor for the growth of a wide variety of gram-positive and gram-negative microorganisms. It is intended for research only. The bottle concentration is 50mg/ml.
Catalog No. NC0753440 Supplier IBI SCIENTIFIC 3P Supplier No. IB02030
Gentamycin Sulfate Solution - 20ml Gentamycin Sulfate Solution is used as an inhibitor for the growth of a wide variety of gram-positive and gram-negative microorganisms. It is intended for research only. The bottle concentration is 50mg/ml.