Learn More
Crescent Chemical Co Inc GENTAMYCIN SULFATE
Supplier: Crescent Chemical Co Inc SE22185.01
Gentamycin sulfate research grade, Ph. Eur. Min. 590 U/mg. Aminoglycoside antibiotic complex from Micromonospora purpurea.Consists of closely related compounds: gentamycin C1, C1a, C2, C2a and C2b; min. 590 mg gentamicin base/g. Inhibits bacterial protein synthesis