Learn More
Sigma Aldrich Fine Chemicals Biosciences Aldrich Essentials beaker, Griffin
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z74057612EA
Encompass
low form, capacity 250 mL, subdivision, 25 mL (graduations: 25-200 mL)
Catalog No. NC2541679