Learn More
Crescent Chemical Co Inc Gentamtcin Sulfate, 1405-41-0, MFCD00270181, 5g
SDP
Supplier:  Crescent Chemical Co Inc SE22185.02
Encompass_Preferred
Molecular Formula C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S, Molecular Weight 1488.79, Research Grade, Min. 590 U/mg. Aminoglycoside antibiotic complex from Micromonospora purpurea.Consists of closely related compounds: gentamycin C1, C1a, C2, C2a and C2b; min. 590 mg gentamicin base/g. Inhibits bacterial protein synthesis by binding to the ribosomal 30S subunit and causing misreading of mRNA (in a similar way as streptomycin). Broad spectrum antibiotic which inhibits growth of many gram positive and gram negative bacteria including strains which are resistant to chloramphenicol, kanamycin or tetracycline. Frequently used in cell culture, often in combination with amphotericin B, nystatin or penicillin G.
Catalog No. 50-247-622