Learn More
AG Scientific Inc Gentamycin Sulfate, 25 G
Supplier:  AG Scientific Inc G106725GM
Encompass_Preferred
Gentamicin Sulfate is an aminoglycoside complex and a broad-spectrum antibiotic created by fermentation of Micromonospora purpurea. It can effectively inhibit disease manifestations produced by Staphylococcus aureus, Klebsiella pneumoniae and other pathogenic organisms. This compound can also inhibit protein synthesis by binding to L6 protein of 50S ribosomal subunit.CAS Number: 1405-41-0Molecular Weight: 575.6 DaChemical Formula: C21H43N5O7 H2SO4Solubility: Soluble in DMSOStorage Temperature: +4CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6917