Learn More
ALADDIN SCIENTIFIC CORPORATION Gentamycin Sulfate, 5g
Catalog No. 5000082545
Encompass_Preferred
Gentamicin sulfate a kind of aminoglycoside antibiotic inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture Gentamicin sulfate inhibits DNase I with an IC50 of 0 57 mM
Catalog No. 50-000-82545 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. G1003915G
Gentamicin sulfate a kind of aminoglycoside antibiotic inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture Gentamicin sulfate inhibits DNase I with an IC50 of 0 57 mM